Orders LinkContactUsLinkComponent
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® - tube with 1 ml SRK® solution, + 1 tube with 1 ml SRK® solution in a sponge + 1 regular FLOQSwabs®, in triple protection wrapping; 10×20 pk; 139 mm choked tube/12×80 mm skirted conic bottom, 1 ml/1 ml, grøn/grøn
Catalog # COPA958C
Supplier:   Copan
SRK® Surfaces and Equipment Microbial Testing Devices
Catalog # COPA958C
Supplier:  Copan
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® - tube with 1 ml SRK® solution, + 1 tube with 1 ml SRK® solution in a sponge + 1 regular FLOQSwabs®, in triple protection wrapping; 10×20 pk; 139 mm choked tube/12×80 mm skirted conic bottom, 1 ml/1 ml, grøn/grøn
Price
4.110,00 kr
Log in for availability
1 * 200 Items
Product Details & Documents
Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.
  • Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula
    Compliance: SRK® kits are designed compliant with ISO 18593:2018, ISO 18593:2004, ISO 17604:2003, and EU directive 2001/471/E.C
    Media choice for enhanced preservation: Choose between an isotonic solution or a wide range of selective and enrichment broths: Half Fraiser, RVS, BPW 1%, TSB, UNS, and Letheen broth
    Sterile sampling frames: the definition of a set sampling area is essential for microbial bioburden validations
    Sampling frames will make your surface sampling match that standard
This product line groups a variety of product codes, each optimised to address specific needs.
Specifications
  • Beskrivelse:
    SRK® - tube with 1 ml SRK® solution, + 1 tube with 1 ml SRK® solution in a sponge + 1 regular FLOQSwabs®, in triple protection wrapping; 10×20 pk; 139 mm choked tube/12×80 mm skirted conic bottom, 1 ml/1 ml
  • Seq:
    31
  • Lågfarve:
    grøn/grøn