Orders LinkContactUsLinkComponent
Stirrer Guides with Anti-clockwise Thread
Stirrer Guides with Anti-clockwise Thread
Stirrer guide, dynamic double seal, 29/32, 10 mm
Katalog # LENZ06306004
Leverandør:   Lenz Laborglas
Stirrer Guides with Anti-clockwise Thread
Katalog # LENZ06306004
Leverandør:  Lenz Laborglas
Stirrer Guides with Anti-clockwise Thread
Stirrer Guides with Anti-clockwise Thread
Stirrer guide, dynamic double seal, 29/32, 10 mm
Price
2.670,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, PFTE/fibre glass pressure screw. The fluoroplastic bearing is in a borosilicate glass housing, and has adjustable PTFE/FPM seal. All parts are interchangeable.
Specifikationer
  • NS:
    29/32
  • Beskrivelse:
    Stirrer guide, dynamic double seal
  • Seq:
    02
  • Skaft - Ø:
    10 mm