Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for AgilePulse™ MAX™

Accessories for AgilePulse™ MAX™

Large volume chamber, 6 mm gap, 6 ml
Catalog # 731-0395
Supplier:   BTX
Accessories for AgilePulse™ MAX™
Catalog # 731-0395
Supplier:  BTX
Accessories for AgilePulse™ MAX™
Accessories for AgilePulse™ MAX™

Large volume chamber, 6 mm gap, 6 ml

Price
2.140,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Large volume chamber, 6 mm gap, 6 ml
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.