Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
5 ml, 70 mm, 0,025 ± ml
  BIBBFRD200
 :   SciLabware
Volumetric flasks, class A, blue markings
  BIBBFRD200
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
5 ml, 70 mm, 0,025 ± ml
409,00 kr
1 * 5 Items
MBL®, borosilicate glass, with smooth neck.
  • Inscriptions in blue enamel
  • Conformity certified, with batch number
ISO 1042
 : ISO 1042
  • Kapacitet:
    5 ml
  • Højde:
    70 mm
  • Tolerance:
    0,025 ± ml