Volumetric flasks, class A, blue markings
5 ml, 70 mm, 0,025 ± ml
BIBBFRD200
MBL®, borosilicate glass, with smooth neck.
- Inscriptions in blue enamel
- Conformity certified, with batch number
ISO 1042
: ISO 1042
- Kapacitet:5 ml
- Højde:70 mm
- Tolerance:0,025 ± ml