Signs, warning stands
Betreten Verboten (German), PP, -
Katalog # BRDY241926
Gå til:
Discontinued
Product Details & Documents
These stable warning devices are easy to transport and suitable for the marking of hazardous zones.
- Made of high-density polypropylene
- Durable, yellow
- No buckling or corrosion
- Large note field is visible from over 9 meters
- With default notices or unlabeled also available
Height: 660 mm
Specifikationer
- Beskrivelse:Betreten Verboten (German)
- Dimensioner:-
- Materiale:PP
- Seq:02