Orders LinkContactUsLinkComponent
Zinc formate 98%
Zinc formate 98%
500 g
  B24210.36
Zinc formate 98%
  B24210.36
 :  557-41-5
Zinc formate 98%
Zinc formate 98%
500 g
 :  557-41-5
2.730,00 kr
1 * 500 g
Purity: 98% CAS No.: 557-41-5 Molecular Formula: C2H2O4Zn Formula Weight: 155.40 UN No.: UN3077 MDL No.: MFCD00050836
  •  :557-41-5
  •  :C₂H₂O₄Zn
  •  :MFCD00050836
  •  :155.40 g/mol
  •  :Stue temperatur
  •  :209-176-5
  •  :9,III
  • Pakn:
    500 g