Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zinc formate 98%

Zinc formate 98%

100 g
Katalog # B24210.22
Irriterende
Leverandør:   Thermo Fisher Scientific
Zinc formate 98%
Katalog # B24210.22
CAS nummer:  557-41-5
Zinc formate 98%
Zinc formate 98%

100 g

CAS nummer:  557-41-5
Price
718,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 557-41-5 Molecular Formula: C2H2O4Zn Formula Weight: 155.40 UN No.: UN3077 MDL No.: MFCD00050836

  • CAS nummer:557-41-5
  • Formel:C₂H₂O₄Zn
  • MDL nummer:MFCD00050836
  • Molecular Weight:155.40 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-176-5
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.