Orders LinkContactUsLinkComponent
Accessories for empty columns HiScale™
Accessories for empty columns HiScale™
Tubing, inner Ø 2 mm, for HiScale™ 50
Katalog # 28-9663-76
Leverandør:   Cytiva
Accessories for empty columns HiScale™
Katalog # 28-9663-76
Leverandør:  Cytiva
Accessories for empty columns HiScale™
Accessories for empty columns HiScale™
Tubing, inner Ø 2 mm, for HiScale™ 50
Price
839,00 kr
Log ind for tilgængelighed
1 * 1 m
Specifikationer
  • Beskrivelse:
    Tubing, inner Ø 2 mm, for HiScale™ 50
  • Seq:
    14