Cleaned foam cartridges
Small XAD cartridge, 30 mm, O.D.×L: 22×100 mm, PU-skum
Katalog # REST22117
Leverandør: Restek
Gå til:
Product Details & Documents
These precleaned cartridges are suitable for the air sampling during ambient, indoor and industrial hygiene applications.
- Ready to use for collection
- Made from cleaned PU foam
- PUF/XAD-2 sandwiches for semi-volatiles
Used in monitoring semi-volatiles organic compounds (SVOC) such as pesticides, PCB, and PAH.
Specifikationer
- Beskrivelse:Small XAD cartridge, 30 mm, O.D.×L: 22×100 mm
- Materiale:PU-skum
- Seq:01



