Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cleaned foam cartridges

Cleaned foam cartridges

Small cartridge, 76 mm, O.D.×L: 22×100 mm, PU-skum
Katalog # REST22116
Leverandør:   Restek
Cleaned foam cartridges
Katalog # REST22116
Leverandør:  Restek
Cleaned foam cartridges
Cleaned foam cartridges

Small cartridge, 76 mm, O.D.×L: 22×100 mm, PU-skum

Price
911,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These precleaned cartridges are suitable for the air sampling during ambient, indoor and industrial hygiene applications.

  • Ready to use for collection
  • Made from cleaned PU foam
  • PUF/XAD-2 sandwiches for semi-volatiles

Used in monitoring semi-volatiles organic compounds (SVOC) such as pesticides, PCB, and PAH.

Specifikationer
  • Beskrivelse:
    Small cartridge, 76 mm, O.D.×L: 22×100 mm
  • Materiale:
    PU-skum
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.