Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Batterier

Batterier

Rechargable NIMH Power ACCU™, Power AA/HR6, 1,2 V, 2600 mAh
Catalog # VART5716101402
Batterier
Catalog # VART5716101402
Batterier
Batterier

Rechargable NIMH Power ACCU™, Power AA/HR6, 1,2 V, 2600 mAh

Price
286,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents

Høj kapacitet med lange driftstider pr. opladning. Kræver ikke en speciel oplader.

  • Kan genoplades op til 1000 gange
Specifikationer
  • Model:
    Power AA/HR6
  • Spænding:
    1,2 V
  • Kapacitet:
    2600 mAh
  • Beskrivelse:
    Rechargable NIMH Power ACCU™
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.