Orders LinkContactUsLinkComponent
VWR®, Büchner funnels
97 mm, 240 ml, 90 mm
Catalog # 511-0058
Supplier:   VWR Collection
VWR®, Büchner funnels
Catalog # 511-0058
Supplier:  VWR Collection
VWR®, Büchner funnels
VWR®, Büchner funnels
97 mm, 240 ml, 90 mm
Price
632,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
White glazed porcelain.
For paper or membrane filtration discs.
Specifications
  • Plade Ø:
    97 mm
  • Kapacitet:
    240 ml
  • Seq:
    06
  • Til filter Ø:
    90 mm