VWR®, Pestles
24 mm, 115 mm
Katalog # 410-0119
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Porcelain.
- Grinding surface unglazed
- Glazed handle
Certifications: Produced according to DIN 12906.
Documents
410-0122
Billeder 360
Specifikationer
- Ø hoved:24 mm
- Længde:115 mm