Orders LinkContactUsLinkComponent
VWR®, Mortars, without Pestles
VWR®, Mortars, without Pestles
140 ml, 50 mm, 85 mm
Katalog # 410-0111
Leverandør:   VWR Collection
VWR®, Mortars, without Pestles
Katalog # 410-0111
Leverandør:  VWR Collection
VWR®, Mortars, without Pestles
VWR®, Mortars, without Pestles
140 ml, 50 mm, 85 mm
Price
208,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Porcelain, produced according to DIN 12906.
  • With spout
  • Glazed outside, grinding surface unglazed
  • Highly resistant to acids and alkalis
Specifikationer
  • Ø indv.:
    85 mm
  • Kapacitet:
    140 ml
  • Højde:
    50 mm