Orders LinkContactUsLinkComponent
VWR®, Mortars, without Pestles
VWR®, Mortars, without Pestles
220 ml, 64 mm, 105 mm
Katalog # 410-0113
Leverandør:   VWR Collection
VWR®, Mortars, without Pestles
Katalog # 410-0113
Leverandør:  VWR Collection
VWR®, Mortars, without Pestles
VWR®, Mortars, without Pestles
220 ml, 64 mm, 105 mm
Price
282,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Porcelain, produced according to DIN 12906.
  • With spout
  • Glazed outside, grinding surface unglazed
  • Highly resistant to acids and alkalis
Specifikationer
  • Ø indv.:
    105 mm
  • Kapacitet:
    220 ml
  • Højde:
    64 mm