VWR®, Mortars, without Pestles
400 ml, 71 mm, 125 mm
Katalog # 410-0114
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Porcelain, produced according to DIN 12906.
- With spout
- Glazed outside, grinding surface unglazed
- Highly resistant to acids and alkalis
Specifikationer
- Ø indv.:125 mm
- Kapacitet:400 ml
- Højde:71 mm