VWR®, Pestles
58 mm, 210 mm
Catalog # 410-0124
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain.
- Grinding surface unglazed
- Glazed handle
Certifications: Produced according to DIN 12906.
Documents
410-0122
Images 360
Specifications
- Ø hoved:58 mm
- Længde:210 mm