VWR®, Pestles
73 mm, 255 mm
Katalog # 410-0125
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Porcelain.
- Grinding surface unglazed
- Glazed handle
Certifications: Produced according to DIN 12906.
Documents
410-0122
Billeder 360
Specifikationer
- Ø hoved:73 mm
- Længde:255 mm