VWR®, Pestles
86 mm, 305 mm
Catalog # 410-0126
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain.
- Grinding surface unglazed
- Glazed handle
Certifications: Produced according to DIN 12906.
Documents
410-0122
Images 360
Specifications
- Ø hoved:86 mm
- Længde:305 mm