Roller bottles, Nunc™ TufRol™
Rulleflasker, TufRol™, glat overflade, med skruelåg uden ventilering, der er nem at sætte på og tage af, 850 cm², 2 pr. pakke, 20 pr. kasse
Catalog # 392-0336
Supplier: Thermo Fisher Scientific
Roller bottles, Nunc™ TufRol™
Catalog # 392-0336
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, sterile.
- One-piece seamless design reduces possibility of leaking through a seam
- Shallow and deep indents allow for excellent manual and automated handling
- Proprietary manufacturing process allows for greater impact resistance
- Vertical expanded surfaces version facilitates emptying and reduces retention of product
Nunc™ TufRol™ have been developed for applications such as the industrial scale production of vaccines, monoclonal antibodies and biotherapeutics, where reliability and reproducibility are essential.
Caution: Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifications
- Dyrkningsareal:850 cm²
- Pakket:2 pr. pakke, 20 pr. kasse
- Beskrivelse:Rulleflasker, TufRol™, glat overflade, med skruelåg uden ventilering, der er nem at sætte på og tage af
- Cat. No.:392-0336
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 392-0326Vented caps for TufRol™ roller bottles, HDPE, sterileLog in for availability1 * 500 Items - 4.110,00 kr