Orders LinkContactUsLinkComponent
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®, 100 mL
Katalog # 1.09032.0100
Leverandør:   Merck
Ion chromatography Multi-Element standards, Supelco®
Katalog # 1.09032.0100
Leverandør:  Merck
CAS nummer:  
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®, 100 mL
CAS nummer:  
Price
2.380,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Beskrivelse:
    Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®
Specifikationer