Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 7940 Series, width 110 mm, BBP™11, 70 m, sort
BRDY804457
: Brady
- Farve:sort
- Beskrivelse:Thermal transfer printer ribbon, 7940 Series, width 110 mm
- Længde:70 m
- Seq:19
- Til:BBP™11