Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Centrifugerør

Tubes, with yellow screw cap, 15 ml, 16×100 mm, Skruelåg, gul, non sterile
Katalog # KART88303/06
Leverandør:   KARTELL
Centrifugerør
Katalog # KART88303/06
Leverandør:  KARTELL
Centrifugerør
Centrifugerør

Tubes, with yellow screw cap, 15 ml, 16×100 mm, Skruelåg, gul, non sterile

Price
1.600,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents

PP with quick closure PE screw cap.

  • Leak-proof
  • Can be centrifuged up to 4000×g
  • Cap is designed to avoid any contamination due to drops of liquid on the inner surface
Specifikationer
  • Lukkestørrelse:
    Skruelåg
  • Ø×L:
    16×100 mm
  • Version:
    non sterile
  • Kapacitet:
    15 ml
  • Beskrivelse:
    Tubes, with yellow screw cap
  • Seq:
    04
  • Lågfarve:
    gul