Accessories for DOSE IT
Silicone tubing, autoclavable, length: 25 m (bulk roll), 8 mm
Catalog # INTG171038
Supplier: INTEGRA Biosciences
Jump to:
Product Details & Documents
Documents
Dose It
Videoer
Specifications
- Beskrivelse:Silicone tubing, autoclavable, length: 25 m (bulk roll)
- Ø indv.:8 mm



