Petri Dishes
Without vents, 120 mm, 14 mm, Aseptisk, 32×10/bag
Leverandør: Anicrin
Gå til:
Product Details & Documents
Suitable for all microbiology applications.
Material: PS
Specifikationer
- Ø:120 mm
- Pakket:32×10/bag
- Sterilitet:Aseptisk
- Version:Without vents
- Højde:14 mm
- Seq:12