Microplates, 96-well, non binding
96-well microplate, F-bottom, non binding, chimney well, μClear® bottom, sort/klar
Katalog # 738-0173
Leverandør: Greiner Bio-One
Microplates, 96-well, non binding
Katalog # 738-0173
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Specifikationer
- Farve:sort/klar
- Beskrivelse:96-well microplate, F-bottom, non binding, chimney well, μClear® bottom
- Seq:07
- Cat. No.:738-0173
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning