Orders LinkContactUsLinkComponent
Separating funnels
Separating funnels
50 ml, 14/23 NS
Katalog # QVFLALG0050701
Leverandør:   QVF LABORTECHNIK
Separating funnels
Katalog # QVFLALG0050701
Leverandør:  QVF LABORTECHNIK
Separating funnels
50 ml, 14/23 NS
Price
925,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Certifications: Complies to DIN 12 451.
Specifikationer
  • Kappestr.:
    14/23 NS
  • Kapacitet:
    50 ml
  • Seq:
    01