Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cleaning brushes for samplers

Nylon, 7 - 14 mm, Wire, 1600 mm
Catalog # 300-0514
Supplier:   Sampling Systems
Cleaning brushes for samplers
Catalog # 300-0514
Supplier:  Sampling Systems
Cleaning brushes for samplers
Cleaning brushes for samplers

Nylon, 7 - 14 mm, Wire, 1600 mm

Price
501,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.

  • Available in a range of lengths and diameters
Specifications
  • Børstemateriale:
    Nylon
  • Børstehoved Ø:
    7 - 14 mm
  • Handle material:
    Wire
  • Seq:
    03
  • Samlet længde:
    1600 mm