Labels for MiniMark printer, type B-7569
Label printer tape, B-7569, Vinyl, Permanent akryl, Thermal transfer, gul, MiniMark™, 57 mm, 57 mm×35 m, 35 m
Katalog # BRDY120855
Leverandør: Brady
Gå til:
Product Details & Documents
The B-7569 vinyl labelling tape is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels.
- Gloss finish
- Permanent acrylic
Documents
Specifikationer
- Printer:MiniMark™
- Printing method:Thermal transfer
- Model:B-7569
- Farve:gul
- Beskrivelse:Label printer tape
- Dimensioner:57 mm×35 m
- Længde:35 m
- Materiale:Vinyl
- Seq:36
- Bredde:57 mm
- Klæbestof:Permanent akryl