Double spatulas, powder
150 mm, 9 mm
Catalog # 231-0545
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel.
- With round handle, one flat and one scoop shaped, rounded spatula end
- Straight shape
- Very good thermal resistance, autoclavable
- Good chemical resistance
Specifications
- Længde:150 mm
- Bladbredde:9 mm