Test sets
Test set I for fumes
Katalog # DRAE8103410
Leverandør: Dräger
Gå til:
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifikationer
- Beskrivelse:Test set I for fumes
- Seq:06
- Cat. No.:DRAE8103410
- Model:Test set I for fumes
- Måletid:180 s