Test sets
Test sets for fumes
Katalog # DRAE8103380
Leverandør: Dräger
Gå til:
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifikationer
- Beskrivelse:Test sets for fumes
- Seq:05
- Cat. No.:DRAE8103380
- Model:Test sets for fumes
- Måletid:240 s