Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant ACY3 (from E. coli)

Human Recombinant ACY3 (from E. coli)

Katalog # ABCAAB109954-250
Marketsource
Leverandør:   Abcam
Human Recombinant ACY3 (from E. coli)
Katalog # ABCAAB109954-250
Leverandør:  Abcam
Marketsource
CAS nummer:  
Human Recombinant ACY3 (from E. coli)
Human Recombinant ACY3 (from E. coli)
CAS nummer:  
Price
17.430,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human ACY3 protein is a Human Full Length protein, in the 1 to 319 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Protein/peptide name:
    ACY3
  • Protein synonyms:
    ASPA2|N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming)|Acylase III|Aminoacylase-3|Aspartoacylase-2|Hepatitis C virus core-binding protein 1|ACY-3|HCBP1|HCV core-binding protein 1
  • Biologisk aktivitet:
    Standard
  • Koncentration:
    0,5 mg/ml
  • Dyrefri:
    N
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB109954-250
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 8, Constituents: 10% Glycerol (glycerin, glycerine), 0,316% Tris HCl, 0,058% Sodium chloride, 0,0154% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Renhed:
    >90%
  • Tested applications:
    MS, SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.