Flathead mop covers and hardware, Slimline™
NovaPoly™ mop cover, irradiated, width 460 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
Katalog # 129-0629
Leverandør: MICRONOVA
Flathead mop covers and hardware, Slimline™
Katalog # 129-0629
Leverandør: MICRONOVA
Product Details & Documents
A series of elasticised slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitising all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
- Covers are easy to change and fit securely over mop frame
- Fully encased Urethane Foam layer for ample disinfectant dispersal
- Single-ply covers available without foam interior
- Fits on QPSL plastic frame or QDSL stainless steel frame
- Suitable for use with strong disinfectants and repeated autoclaving
The Micronova Slimline™ Flat Head Mop Covers fit easily and securely onto the 360° swivel SlimLine™ plastic or stainless steel mop adapters, which can be locked at 180° for more controlled cleaning.
Bestillingsoplysninger: Alle komponenter skal bestilles separat.
Documents
MICRSLSC6-14_TDS_EN
Datablad
Specifikationer
- Beskrivelse:NovaPoly™ mop cover, irradiated, width 460 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
- Seq:06
- Cat. No.:129-0629
- Environmentally Preferable:Reduktion af affald
Complementary Products
Produktinformation
- Katalog #BeskrivelseMaterialeLængdeTilgængelighedPrice
- 129-0551Håndtag, teleskopisk, rustfrit stål, Længde 965-1830 mmRustfrit stål 304965 - 1829 mmLog ind for tilgængelighed1 * 1 Items - 2.870,00 kr
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)129-0537Log ind for tilgængelighed1 * 1 Items - 3.470,00 kr
Produktinformation
- Katalog #L×BTilgængelighedPrice
- SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handlesSlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handlesMICRQPSL-099×4" (22.9×10 cm) mmLog ind for tilgængelighed1 * 1 Items - 1.210,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- MicroMop® Adapter
Produktinformation
- Katalog #L×BTilgængelighedPrice
- SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handlesSlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handles129-097614×4" (35.5×10 cm) mmLog ind for tilgængelighed1 * 1 Items - 1.320,00 kr
Produktinformation
- Katalog #L×BTilgængelighedPrice
- SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handlesSlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handlesMICRQPSL-1818×4" (45.7×10 cm) mmLog ind for tilgængelighed1 * 1 Items - 1.450,00 kr
Produktinformation
- Katalog #L×BTilgængelighedPrice
- SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handlesSlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles129-053818×4" (45.7×10 cm) mmLog ind for tilgængelighed1 * 1 Items - 3.610,00 kr
Produktinformation
- Katalog #L×BTilgængelighedPrice
- SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handlesSlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles129-05369×4" (22.9×10 cm) mmLog ind for tilgængelighed1 * 1 Items - 3.050,00 kr
Produktinformation
- Katalog #L×BTilgængelighedPrice
- SlimLine™ plastic mop adapters, polypropylene and polyketone material, use with 16" flat head mopsSlimLine™ plastic mop adapters, polypropylene and polyketone material, use with 16" flat head mopsMICRQPSL-16410×100 mmLog ind for tilgængelighed1 * 1 Items - 1.380,00 kr