Orders LinkContactUsLinkComponent
Flathead mop covers and hardware, Slimline™
NovaPoly™ mop cover, irradiated, width 460 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
Katalog # 129-0629
   Reduktion af affald
Leverandør:   MICRONOVA
Flathead mop covers and hardware, Slimline™
Katalog # 129-0629
Leverandør:  MICRONOVA
   Reduktion af affald
Flathead mop covers and hardware, Slimline™
Flathead mop covers and hardware, Slimline™
NovaPoly™ mop cover, irradiated, width 460 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
Price
4.350,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
A series of elasticised slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitising all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
  • Covers are easy to change and fit securely over mop frame
  • Fully encased Urethane Foam layer for ample disinfectant dispersal
  • Single-ply covers available without foam interior
  • Fits on QPSL plastic frame or QDSL stainless steel frame
  • Suitable for use with strong disinfectants and repeated autoclaving
The Micronova Slimline™ Flat Head Mop Covers fit easily and securely onto the 360° swivel SlimLine™ plastic or stainless steel mop adapters, which can be locked at 180° for more controlled cleaning.
 : Alle komponenter skal bestilles separat.
Specifikationer
  • Beskrivelse:
    NovaPoly™ mop cover, irradiated, width 460 mm, NovaPoly™ 100% Smooth Knit Polyester (Urethane foam interior)
  • Seq:
    06
  • Cat. No.:
    129-0629
  • Environmentally Preferable:
    Reduktion af affald
Complementary Products
Produktinformation
  • Katalog #
    Beskrivelse
    Materiale
    Længde
    Tilgængelighed
    Price
  • 129-0551
    Håndtag, teleskopisk, rustfrit stål, Længde 965-1830 mm
    Rustfrit stål 304
    965 - 1829 mm
    Log ind for tilgængelighed
    1 * 1 Items - 2.870,00 kr
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 14" flat head mops; SSU series handles, 14×4" (35.5×10 cm)
    129-0537
    Log ind for tilgængelighed
    1 * 1 Items - 3.470,00 kr
Produktinformation
  • Katalog #
    L×B
    Tilgængelighed
    Price
  • SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handles
    SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 9" flat head mops; STA series handles , 9" flat head mops; STA series handles
    MICRQPSL-09
    9×4" (22.9×10 cm) mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.210,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • MicroMop® Adapter
    MicroMop® Adapter
    129-0170
    Log ind for tilgængelighed
    1 * 1 Items - 1.260,00 kr
Produktinformation
  • Katalog #
    L×B
    Tilgængelighed
    Price
  • SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handles
    SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 14" flat head mops; STA series handles
    129-0976
    14×4" (35.5×10 cm) mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.320,00 kr
Produktinformation
  • Katalog #
    L×B
    Tilgængelighed
    Price
  • SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handles
    SlimLine™ Plastic mop frame, 360° swivel, ACME connect, 18" flat head mops; STA series handles
    MICRQPSL-18
    18×4" (45.7×10 cm) mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.450,00 kr
Produktinformation
  • Katalog #
    L×B
    Tilgængelighed
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 18" flat head mops; SSU series handles
    129-0538
    18×4" (45.7×10 cm) mm
    Log ind for tilgængelighed
    1 * 1 Items - 3.610,00 kr
Produktinformation
  • Katalog #
    L×B
    Tilgængelighed
    Price
  • SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles
    SlimLine™ Stainless steel mop frame, 360° swivel, quick disconnect, 9" flat head mops; SSU series handles
    129-0536
    9×4" (22.9×10 cm) mm
    Log ind for tilgængelighed
    1 * 1 Items - 3.050,00 kr
Produktinformation
  • Katalog #
    L×B
    Tilgængelighed
    Price
  • SlimLine™ plastic mop adapters, polypropylene and polyketone material, use with 16" flat head mops
    SlimLine™ plastic mop adapters, polypropylene and polyketone material, use with 16" flat head mops
    MICRQPSL-16
    410×100 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.380,00 kr