Orders LinkContactUsLinkComponent
Cryoboxes, Cardboard
7×7, 17 mm
  HERA820049
Cryoboxes, Cardboard
  HERA820049
Cryoboxes, Cardboard
Cryoboxes, Cardboard
7×7, 17 mm
36,60 kr
1 * 1 Items
Cryo resistant cardboard.
  • Use with cardboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
  • Rum:
    7×7
  • Til rør Ø:
    17 mm
  • Seq:
    07