Orders LinkContactUsLinkComponent
Cryoboxes, Cardboard
Cryoboxes, Cardboard
10×10, 12 mm
Katalog # HERA820100
Leverandør:   Thermo Scientific
Cryoboxes, Cardboard
Katalog # HERA820100
Leverandør:  Thermo Scientific
Cryoboxes, Cardboard
Cryoboxes, Cardboard
10×10, 12 mm
Price
36,60 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cryo resistant cardboard.
  • Use with cardboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
Documents
Specifikationer
  • Rum:
    10×10
  • Til rør Ø:
    12 mm
  • Seq:
    11