Cryoboxes, Cardboard
80 mm Cryobox with 81-cell divider, square shaped, 0,08 kg, 9×9, frysere med ultralav temperatur, hvid, 127×127×80 mm
Katalog # HERA820003
Leverandør: Thermo Scientific
Gå til:
Product Details & Documents
Cryo resistant cardboard.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
Documents
HERA820002_TDS_EN
Datablad
Specifikationer
- Rum:9×9
- Til:frysere med ultralav temperatur
- B×D×H udv.:127×127×80 mm
- Farve:hvid
- Beskrivelse:80 mm Cryobox with 81-cell divider, square shaped, 0,08 kg
- Seq:04