Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 4900 series, width 65 mm, BBP™11, 70 m, sort
Katalog # BRDY804793
Leverandør: Brady
Gå til:
Product Details & Documents
Specifikationer
- Farve:sort
- Beskrivelse:Thermal transfer printer ribbon, 4900 series, width 65 mm
- Længde:70 m
- Seq:06
- Til:BBP™11