Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Organic reference standards, Formaldehyde IC Standards, Supelco®

Formaldehyde IC Standard, 1000 μg/ml in H₂O, Analytical standard, 100 mL
Katalog # ICS045-100ML
Leverandør:   Merck
Organic reference standards, Formaldehyde IC Standards, Supelco®
Katalog # ICS045-100ML
Leverandør:  Merck
CAS nummer:  
Organic reference standards, Formaldehyde IC Standards, Supelco®

Formaldehyde IC Standard, 1000 μg/ml in H₂O, Analytical standard, 100 mL

CAS nummer:  
Price
849,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

 : This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.

 

Specifikationer

Specifikationer

  • Koncentration:
    1000 μg/ml in H₂O
  • Pakn:
    100 mL
  • Standard:
    Analytical standard
  • Beskrivelse:
    Formaldehyde IC Standard
  • Cat. No.:
    ICS045-100ML
  • Beskrivelse:
    Formaldehyde IC Standard

Specifikationer