Sprøjtedispenser, Microlab® 600
Product Details & Documents
The Microlab® 600 dispensers are highly precise syringe pumps with a touchscreen graphical user interface designed to quickly and easily dispense liquids. This positive displacement system provides better than 99% accuracy, independent of liquid viscosity, vapour pressure and temperature. The inert fluid path minimises sample carry over and improves compatibility with harsh chemicals. There are three different dispenser models available.
- Dispenserer volumener fra 100 nl til 50 ml
- Stabil væskevej, der kan anvendes til barske kemikalier
The single syringe dispenser is ideal for dedicated dispensing applications requiring a single syringe drive. The dual syringe dispenser has two syringe drives for applications such as animal dosing that require the use of more than one liquid at the same time. The dual syringe continuous dispenser is designed for time critical applications requiring repetitive dispensing.
The basic controller comes standard with a quick start run screen that provides basic diluting and dispensing functionality. The advanced controller delivers additional functions through intuitive wizards and the custom method editor. Wizard settings can be saved as favorites or custom methods on the hardware key. The basic controller can be upgraded to the advanced controller if it is not selected initially.
The standalone syringe pump offers total control with customised program methods using Visual C#®, Visual Basic®, and LabVIEW®. Commands can be deployed to any instrument on the network from anywhere in the world. The API is compatible with Windows® computers running Microsoft®. NET version 2.0 or higher.
Certifications: CE marked. CSA certified.
Accessories information: The Bubble Free Prime (BFP) syringe is specifically designed for the Microlab® 600 series and the only syringe type compatible with the instrument. It has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Ordering information: Dispensers are supplied with a Concorde hand probe (dual dispensers use the dual push button hand probe), universal valves, fill/dispense tubing, accessory holder, country specific power cord and default syringes. Standalone pumps are supplied with universal valves, country specific power cord and default syringes. To order a dispenser or standalone pump with other syringe sizes, please contact your local Avantor sales office.
Specifikationer
- Beskrivelse:Dual syringe dispenser with advanced controller, Microlab 625-DIS, two 1 ml BFP default syringes
- Seq:04
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Luer lock conversion kit
Produktinformation
- Katalog #TilgængelighedPrice
- Dispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPDispense tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPHAMI1171-02Log ind for tilgængelighed1 * 1 Items - 648,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Dispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPDispense tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPHAMI1173-02Log ind for tilgængelighed1 * 1 Items - 648,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Screen protector, PET
Produktinformation
- Katalog #TilgængelighedPrice
- Universal power supply, 110 - 220 VACUniversal power supply, 110 - 220 VACHAMI61092-01Log ind for tilgængelighed1 * 1 Items - 1.100,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Cable management system for disposable tip hand probeCable management system for disposable tip hand probeHAMI65160-01Log ind for tilgængelighed1 * 1 Items - 1.390,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Airshield for fresh air inside the instrumentAirshield for fresh air inside the instrumentHAMI68562-01Log ind for tilgængelighed1 * 1 Items - 3.840,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Håndprobe med dobbelt trykknap
Produktinformation
- Katalog #TilgængelighedPrice
- Fill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPFill tubing, volume 3,14 µl/mm, custom length, 12 gauge, FEPHAMI1172-02Log ind for tilgængelighed1 * 1 Items - 532,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Fill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPFill tubing, volume 0,79 µl/mm, custom length, 18 gauge, FEPHAMI1174-02Log ind for tilgængelighed1 * 1 Items - 532,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Continuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEPContinuous fill tubing, volume 3,14 µl/mm, 12 gauge, FEPHAMI61491-01Log ind for tilgængelighed1 * 1 Items - 805,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- New Microlab 600 controller, includes SD card slot, serial port, large and small USB portNew Microlab 600 controller, includes SD card slot, serial port, large and small USB portHAMI61500-05Log ind for tilgængelighed1 * 1 Items - 24.320,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Continuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEPContinuous fill tubing, volume 0,79 µl/mm, 18 gauge, FEPHAMI61491-02Log ind for tilgængelighed1 * 1 Items - 714,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Printer kit includes printer, power source, adapter and cablePrinter kit includes printer, power source, adapter and cable613-5822Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Disposable tips for 613-1625, 5 ml
Produktinformation
- Katalog #TilgængelighedPrice
- Fodkontakt til ML 600-serien
Produktinformation
- Katalog #TilgængelighedPrice
- Valve plug
Produktinformation
- Katalog #TilgængelighedPrice
- Håndprobe med engangsspids, stort volumen (1 - 5 ml)Håndprobe med engangsspids, stort volumen (1 - 5 ml)613-1625Log ind for tilgængelighed1 * 1 Items - 13.810,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Probe holder and tubing stand
Produktinformation
- Katalog #TilgængelighedPrice
- Fill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEPFill tubing, volume 0,96 ml, 1219 mm, 18 gauge, FEP613-1582Log ind for tilgængelighed1 * 1 Items - 391,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Fill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEPFill tubing, volume 4,57 ml, 1219 mm, 12 gauge, FEP613-1583Log ind for tilgængelighed1 * 1 Items - 459,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)Custom programmer kit (compatible with Microsoft® .NET 2.0 and higher)HAMI61500-03Log ind for tilgængelighed1 * 1 Items - 2.590,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Concorde CT håndprobe
Produktinformation
- Katalog #TilgængelighedPrice
- Valve cross tube assembly
Produktinformation
- Katalog #TilgængelighedPrice
- Venstremontering
Produktinformation
- Katalog #TilgængelighedPrice
- Dispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEPDispense tubing, volume 1,08 ml, 1372 mm, 18 gauge, FEP613-1584Log ind for tilgængelighed1 * 1 Items - 399,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Dispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEPDispense tubing, volume 5,15 ml, 1372 mm, 6,35 - 711,2 mm UNF, 12 gauge, FEP613-1585Log ind for tilgængelighed1 * 1 Items - 488,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Højremontering
Produktinformation
- Katalog #TilgængelighedPrice
- Disposable tip hand probe, 0,5 - 1000 µlDisposable tip hand probe, 0,5 - 1000 µl613-5823Log ind for tilgængelighed1 * 1 Items - 6.620,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Tubing clips
Produktinformation
- Katalog #TilgængelighedPrice
- Basic to Advanced Controller upgrade kit, including 2 GB SD cardBasic to Advanced Controller upgrade kit, including 2 GB SD cardHAMI61500-02Log ind for tilgængelighed1 * 1 Items - 6.440,00 kr