Orders LinkContactUsLinkComponent
Supports for stand grids
flade flanger LF 200, strukturgitter 0,500 m
Katalog # LENZ06247520
Leverandør:   Lenz Laborglas
Supports for stand grids
Katalog # LENZ06247520
Leverandør:  Lenz Laborglas
Supports for stand grids
Supports for stand grids
flade flanger LF 200, strukturgitter 0,500 m
Price
18.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel (clamp), steel, powder coated (supports).
For fixing lids onto reaction vessels and for the suspension into stands, complete with clamp and supports.
Specifikationer
  • Til:
    flade flanger LF 200, strukturgitter 0,500 m
  • Seq:
    03