Orders LinkContactUsLinkComponent
VWR®, Agarose I™
Agarose I™
  0710-250G
VWR®, Agarose I™
  0710-250G
VWR®, Agarose I™
VWR®, Agarose I™
Agarose I™
4.240,00 kr
1 * 250 g
Agarose I™ is a standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications (250 bp to 22 kb).
  • All-purpose, high purity agarose
  • Exceptional band resolution and clarity
  • Nuclease- and protease-free
Agarose I™ has a low EEO (electroendosmosis) for shorter electrophoretic runs without compromising resolution. Agarose I™ is excellent for blotting techniques and general manipulations.
  • Beskrivelse:
    Agarose I™
  • Seq:
    03