Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Vand for HPLC

Vand for HPLC

4 L
Katalog # K978-4L
   Sikrere sundhed for mennesker og miljø
Leverandør:   VWR Chemicals
Vand for HPLC
Katalog # K978-4L
Leverandør:  VWR Chemicals
   Sikrere sundhed for mennesker og miljø
CAS nummer:  7732-18-5
Vand for HPLC
Vand for HPLC

4 L

CAS nummer:  7732-18-5
Price
2.390,00 kr
Log ind for tilgængelighed
1 * 4 L
Product Details & Documents

High purity water.

  • Functionally tested reagents suitable for HPLC and mass spectrometry
  • Low UV absorbing and low particulate count to reduce background
Synonymer:  Hydrogen oxide
  • CAS nummer:7732-18-5
  • Formel:H₂O
  • MDL nummer:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Kogende Pt:100 °C (1013 hPa)
  • Smeltende Pt:0 °C
  • Tæthed:1 g/cm³ (4 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-791-2
Specifikationer

Specifikationstestresultater

  • Abs.@200nm:
    0.1
  • Abs.@210nm:
    0.1
  • Abs.@254nm:
    0.005
  • Abs.@300nm:
    0.005
  • Ammonia:
    0.3 ppm
  • Calcium Test:
    Pass
  • Carbon Dioxide Test:
    Pass
  • Chloride:
    0.5 ppm
  • DNase:
    none detected
  • Endotoxins:
    0.25 EU/mL
  • Liquid Chromatography Suitability:
    Pass
  • Oxidizable Substances:
    Pass
  • pH @25°C:
    5.0-7.0
  • Protease:
    none detected
  • RNase:
    none detected

Specifikationer

  • Pakn:
    4 L
  • Cat. No.:
    K978-4L
  • Miljøcertificering:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Sikrere sundhed for mennesker og miljø

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.