Jern, powder
Glass bottle for solids, 1.000 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:7439-89-6
- Formel:Fe
- MDL nummer:MFCD00010999
- Molecular Weight:55.85 g/mol
- Kogende Pt:3000 °C (1013 hPa)
- Smeltende Pt:1536 °C
- Tæthed:7,86 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-096-4
- FN:3089
- ADR:4.1,III
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1.000 g



