Orders LinkContactUsLinkComponent
Pipettespidser, universal, Axygen®
Pipettespidser, universal, Axygen®
Bevelled, wide bore (1,5 mm), 1 - 200 µl, +, Transparent, 10 racks, 96 each
Katalog # 615-0029
Leverandør:   Corning
Pipettespidser, universal, Axygen®
Katalog # 615-0029
Leverandør:  Corning
Pipettespidser, universal, Axygen®
Pipettespidser, universal, Axygen®
Bevelled, wide bore (1,5 mm), 1 - 200 µl, +, Transparent, 10 racks, 96 each
Price
550,00 kr
Log ind for tilgængelighed
1 * 960 Items
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
  • PP, autoclavable
  • Smooth finish for minimum retention of collected sample
  • Certified free of RNase, DNase and endotoxins
  • Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Ordering information: Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
  • Pakket:
    10 racks, 96 each
  • Steril:
    +
  • Volumen:
    1 - 200 µl
  • Seq:
    25
  • Spidstype:
    Bevelled, wide bore (1,5 mm)
  • Spits farge:
    Transparent