Pipettespidser, universal, Axygen®
Bevelled, wide bore (1,5 mm), 1 - 200 µl, +, Transparent, 10 racks, 96 each
Katalog # 615-0029
Leverandør: Corning
Gå til:
Product Details & Documents
Disse universelle pipettespidser er Kompatibel med de fleste mikropipetter.
- PP, autoclavable
- Smooth finish for minimum retention of collected sample
- Certified free of RNase, DNase and endotoxins
- Maxymum Recovery® tips feature ultra smooth surfaces: For reduced sample loss and maximal accuracy and precision
Ordering information: Fås løse eller i racks, sterile eller usterile versioner.
Specifikationer
- Pakket:10 racks, 96 each
- Steril:+
- Volumen:1 - 200 µl
- Seq:25
- Spidstype:Bevelled, wide bore (1,5 mm)
- Spits farge:Transparent