Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
mFreSR™ Cryopreservation Medium

mFreSR™ Cryopreservation Medium

mFreSR™, 50 ml bottle, Human; Serum-free, 50 mL
Katalog # STMC05855
Marketsource
Leverandør:   Stemcell Technologies
mFreSR™ Cryopreservation Medium
Katalog # STMC05855
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
mFreSR™ Cryopreservation Medium
mFreSR™ Cryopreservation Medium

mFreSR™, 50 ml bottle, Human; Serum-free, 50 mL

CAS nummer:  
Price
3.058,00 kr
Log ind for tilgængelighed
1 * 50 mL
Product Details & Documents

Serum-free cryopreservation medium for human ES and iPS cells.

  • Easy to use
  • Serum-free formulation, optimised for use with TeSR™ maintenance media
  • High thawing efficiencies
  • Pre-screened components to ensure batch-to-batch consistency

mFreSR™ is a serum-free cryopreservation medium designed for the cryopreservation of human embryonic and induced pluripotent stem cells (ES and iPS cells). mFreSR™ contains DMSO and is complete and ready-to-use. Together with mTeSR™1 or mTeSR™ Plus, mFreSR™ eliminates the use of feeders and serum. Human ES and iPS cells cryopreserved in mFreSR™ have thawing efficiencies higher than reported conventional thawing methods using serum.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    50 mL
  • Beskrivelse:
    mFreSR™, 50 ml bottle
  • Udformning:
    Human; Serum-free

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.