Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Industriøser, plastilab®

Industriøser, plastilab®

1500 ml, 400 mm
Katalog # KART1112
Leverandør:   KARTELL
Industriøser, plastilab®
Katalog # KART1112
Leverandør:  KARTELL
Industriøser, plastilab®
Industriøser, plastilab®

1500 ml, 400 mm

Price
1.480,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents

These scoops made from HDPE are designed for heavy industrial use.

  • Round shaped design facilitates complete transfer of sampling material
  • Suitable for foodstuff
Specifikationer
  • Kapacitet:
    1500 ml
  • Længde:
    400 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.