Orders LinkContactUsLinkComponent
Industriøser, plastilab®
500 ml, 300 mm
  KART1111
 :   KARTELL
Industriøser, plastilab®
  KART1111
 :  KARTELL
Industriøser, plastilab®
Industriøser, plastilab®
500 ml, 300 mm
958,00 kr
1 * 25 Items
These scoops made from HDPE are designed for heavy industrial use.
  • Round shaped design facilitates complete transfer of sampling material
  • Suitable for foodstuff
  • Kapacitet:
    500 ml
  • Længde:
    300 mm
  • Seq:
    02