Orders LinkContactUsLinkComponent
Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
22, 32 mm, sort
Katalog # BDAM368651
Leverandør:   BD Medical
Blood collection needles, BD Vacutainer® Eclipse™
Katalog # BDAM368651
Leverandør:  BD Medical
Blood collection needles, BD Vacutainer® Eclipse™
Blood collection needles, BD Vacutainer® Eclipse™
22, 32 mm, sort
Price
797,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
The BD Vacutainer® Eclipse™ blood collection needle is a safety-engineered, multi-sample blood collection needle that offers a simple, effective way to collect blood while reducing the possibility of needlestick injuries. It features a safety shield that allows for one-handed activation to cover the needle immediately upon withdrawal from the vein and confirms proper activation with an audible click.
  • Built-in safety shield is aligned with the bevel of the needle
  • The safety shield permanently locks into place, significantly reducing the risk of needlestick injuries
  • The pre-attached holder (only supplied with some models) minimises the risk of accidental needlestick injuries from the holder end of the device
Specifikationer
  • Farvekode:
    sort
  • Måler:
    22
  • Længde:
    32 mm
  • Seq:
    04